missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution (1N/Certified), Fisher Chemical™

Catalog No. p-214314
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SS2661 1 L Poly Bottle
SS2664 4 L Poly Bottle
SS26620 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SS2661 Supplier Thermo Fisher Scientific Supplier No. BPSS2661
Only null left

Volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 13.3
Color Colorless
Boiling Point 100°C
Melting Point 0°C
Concentration 0.995 to 1.005N
Quantity 1 L
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide Solution, 1N
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes.
NFPA
Health:3 Flammability:0 Instability:0
DOTInformation DOT Class 8, : Corrosive