missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution (10N/Certified), Fisher Chemical™

Catalog No. p-214277
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Poly Bottle
PolyPac
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SS2551 1 L Poly Bottle
SS2554 4 L Poly Bottle
SS25520 20 L PolyPac
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SS2551 Supplier Thermo Fisher Scientific Supplier No. FLSS2551
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 12.0
Boiling Point >100°C
Melting Point -10°C
Concentration 9.95 to 10.05
UN Number 1824
Density 1.32g/mL
Quantity 1 L
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Vapor Pressure 14mmHg
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide Solution, 10N
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive