missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution (0.02N (N/50)/Certified), Fisher Chemical™

Catalog No. SS2824
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SS2824 4 L Poly Bottle
SS2821 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SS2824 Supplier Thermo Fisher Scientific Supplier No. FLSS2824

A volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Concentration 0.0198 to 0.0202N
Quantity 4 L
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide Solution, 0.02N (N/50)
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive