missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide-Sodium Thiosulfate, For Nitrogen (Organic), Certified, 12.5N ±1.0N, LabChem™
$81.53 - $515.60
Specifications
| Chemical Name or Material | Sodium Hydroxide-Sodium Thiosulfate |
|---|---|
| Color | Colorless |
| Name Note | For Nitrogen (Organic) |
| CAS Max % | 61.5,36,2.5 |
| CAS | 1310-73-2,10102-17-7 |
Specifications
| Sodium Hydroxide-Sodium Thiosulfate | |
| For Nitrogen (Organic) | |
| 1310-73-2,10102-17-7 | |
| 12.5N ±1.0N | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| 40 | |
| Passes Test | |
| Traceable to NIST | |
| Sodium oxide; Thermal decomposition generates: Corrosive vapors; Sulfur compounds |
| Colorless | |
| 61.5,36,2.5 | |
| NaOH | |
| 1.39g/mL | |
| Soluble in water | |
| Certified | |
| 1.39g/mL | |
| 15cSt | |
| Liquid |