missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide-Sodium Thiosulfate, For Nitrogen (Organic), Certified, 12.5N ±1.0N, LabChem™

Supplier:  LabChem LC246002

 View more versions of this product

Catalog No. LC246002

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide-Sodium Thiosulfate
For Nitrogen (Organic)
1310-73-2,10102-17-7
12.5N ±1.0N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
40
Passes Test
1.39g/mL
15cSt
1 L
Colorless
61.5,36,2.5
NaOH
1.39g/mL
Soluble in water
Certified
Poly Bottle
Traceable to NIST
Sodium oxide; Thermal decomposition generates: Corrosive vapors; Sulfur compounds
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions