missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Low Chloride, Pellets, ACS, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S12951KG

 Afficher plus de versions de ce produit

Numéro de catalogue. 18613644


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

1310-73-2
HNaO
1 kg
sodium hydroxide
97%
To Pass Test
1
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]
40.00
ACS
Poly Bottle

Identifiants chimiques

1310-73-2
40.00
sodium hydroxide
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.