missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Low Chloride, Pellets, ACS, 97%, Spectrum™ Chemical

Catalog No. p-7166322
Change view
Click to view available options
Quantity:
125 g
500 g
1 kg
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613642 125 g Poly Bottle
18613643 500 g Poly Bottle
18613644 1 kg Poly Bottle
18613645 2.5 kg Poly Bottle
18613646 12 kg Poly Pail
18613647 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18613642 Supplier Spectrum Chemical Mfg Cor Supplier No. S1295125GM
Only null left

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 97%
Grade ACS
Identification To Pass Test
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.