missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified Reagent Grade, 50.0% (w/w) ±1.0%, LabChem™

$56.33 - $5204.50

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC241501
View Documents
LabChem
LC241501
500 mL
Each for $56.33
Only null left
 
LC241502
View Documents
LabChem
LC241502
1 L
Each for $130.93
Only null left
 
LC241504
View Documents
LabChem
LC241504
4 L
Each for $203.93
Only null left
 
LC241506
View Documents
LabChem
LC241506
10 L
Each for $507.87
Only null left
 
LC241505
View Documents
LabChem
LC241505
20 L
Each for $804.33
Only null left
 
LC24150200
View Documents
LabChem
LC24150200
200 L
Each for $5,204.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide

Specifications

Colorless
1310-73-2,7732-18-5
50,50
NaOH
1,525kg/m3
Soluble in water
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Certified Reagent
Poly Bottle
Traceable to NIST
Sodium Hydroxide
143°C
12°C
HNaO
UN1824
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
500 mL
[OH-].[Na+]
39.997
CHEBI:32145
50.0% ±1.0% (w/w)
Passes Test
1525kg/m3
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 215-185-5

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'