missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified Reagent Grade, 50.0% (w/w) ±1.0%, LabChem™

Supplier:  LabChem LC241505

 View more versions of this product

Catalog No. LC241505


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide
143°C
12°C
HNaO
UN1824
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
20 L
[OH-].[Na+]
39.997
CHEBI:32145
50.0% ±1.0% (w/w)
Passes Test
1525kg/m3
Liquid
Colorless
1310-73-2,7732-18-5
50,50
NaOH
1,525kg/m3
Soluble in water
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Certified Reagent
Poly Drum
Traceable to NIST

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 215-185-5

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions