missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified, 18.0% (w/w) ±0.5% (w/w), 5.39N (5.39M), LabChem™

Catalog No. LC240852
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
LC240852 1 L
1 options

Catalog No. LC240852

Supplier: LabChem LC240852

Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Max % 82, 18
Linear Formula NaOH
Concentration 5.39N (5.39M)
UN Number UN1824
Density 1.2g/mL
Solubility Information Soluble in water
Quantity 1 L
Formula Weight 40
Percent Purity 18.0% ±0.5% (w/w)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.2g/mL
Traceability to NIST Traceable to NIST
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature