missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Certified, 1.000N ±0.005N (1.0M), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 96.16, 3.84 |
| Linear Formula | NaOH |
| Concentration | 1.000N ± 0.005N |
| UN Number | UN1824 |
| Density | 1.04g/mL |
| Solubility Information | Soluble in water |
| Quantity | 20 L |
| Formula Weight | 40 |
| Grade | Certified |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves/eye protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/docto |
| Recommended Storage | Room Temperature |