missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified, 0.1000N ±0.0005N (0.1M), LabChem™

Catalog No. LC242701
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC242701 500 mL Poly Bottle
LC242702 1 L Poly Bottle
LC242704 4 L Poly Bottle
LC242706 10 L Cubitainer™
LC242705 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC242701 Supplier LabChem Supplier No. LC242701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Max % 99.6, 0.4
Linear Formula NaOH
Concentration 0.1000N ±0.0005N (0.1M)
Density 1g/mL
Solubility Information Soluble in water
Quantity 500 mL
Formula Weight 40
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature