missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Certified, 0.0227N ±0.0002N (0.0227M), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 99.91, 0.09 |
| Linear Formula | NaOH |
| Concentration | 0.0227N ±0.0002N (0.0227M) |
| Density | 1g/mL |
| Solubility Information | Soluble in water |
| Quantity | 20 L |
| Formula Weight | 40 |
| Grade | Certified |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |