missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, ACS Grade, > 97.0%, LabChem™
Supplier: LabChem LC239002
Specifications
| Sodium Hydroxide | |
| 1388°C | |
| 323°C | |
| HNaO | |
| UN1823 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| 1 kg | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| >97.0% | |
| Passes Test | |
| 2130kg/m3 |
| White | |
| 1310-73-2 | |
| 100 | |
| NaOH | |
| 2130kg/m3 | |
| Soluble in water | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40 | |
| ACS | |
| Poly Bottle | |
| Pellet |
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life.
GHS P Statement
Do not breathe dust, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Danger
EINECSNumber : 215-185-5
Recommended Storage : Room Temperature