missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, ACS Grade, > 97.0%, LabChem™

Catalog No. p-5379590
Change view
Click to view available options
Quantity:
500 g
1 kg
Catalog No. Quantity
LC239001 500 g
LC239002 1 kg
2 options

Catalog No. LC239001

Supplier: LabChem LC239001

Only null left

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color White
Boiling Point 1,388°C
Melting Point 323°C
CAS Max % 100
Linear Formula NaOH
UN Number UN1823
Density 2,130kg/m3
Solubility Information Soluble in water
Quantity 500 g
Formula Weight 40
Percent Purity >97.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 2130kg/m3
Physical Form Pellet
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe dust, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with applicable regulations.
EINECSNumber 215-185-5
Recommended Storage Room Temperature