missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, ≥98%, Honeywell
Reagent Grade, pellets (anhydrous),≥98%
$219.03 - $1758.00
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 39.997 |
| MDL Number | MFCD00003548 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6001422
|
Honeywell Chemicals
S5881500G |
500 g |
Each for $219.03
|
|
|||||
|
6001421
|
Honeywell Chemicals
S58811KG |
1 kg |
Each for $339.03
|
|
|||||
|
6001423
|
Honeywell Chemicals
S58815KG |
5 kg |
Each for $1,030.24
|
|
|||||
|
60048166
|
Honeywell Chemicals
S58816X1KG |
6 x 1 kg |
Case of 6 Each for $1,758.00
|
|
|||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
Specifications
| 1310-73-2 | |
| NaOH | |
| UN1823 | |
| 500 g | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| ≥98% | |
| 3 mmHg (37°C), 18 mmHg (20°C) |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40g/mol | |
| >1 (vs air) | |
| Sodium hydroxide |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
RTECSNumber : WB4900000