missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 8% (w/v) (80 g/L), Ricca Chemical

Catalog No. p-2917136
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
725616 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 725616 Supplier Ricca Chemical Company Supplier No. 725616
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 500 mL
Percent Purity 7.96 to 8.04%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 8% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide