missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/w) Solution, Technical, Spectrum™ Chemical

Catalog No. 18613622
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
55 gal.
Packaging:
Drum
Poly Bottle
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613622 1 L Poly Bottle
18613623 4 L Poly Bottle
18613624 10 L Poly Drum
18613625 20 L Poly Drum
18613626 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613622 Supplier Spectrum Chemical Mfg Cor Supplier No. S6221LT
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5

Specifications

Concentration 50%
Quantity 1 L
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Sodium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.