missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/w), Solution, 48-52%, Spectrum™ Chemical

Catalog No. 18613621
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
2.5 gal.
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613621 200 L Drum
18613615 100 mL Poly Bottle
18613616 500 mL Poly Bottle
18613617 1 L Poly Bottle
18613618 4 L Poly Bottle
18613620 20 L Poly Drum
18613619 2.5 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18613621 Supplier Spectrum Chemical Mfg Cor Supplier No. S1310200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1310-73-2 | 40.00 g/mol

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5

Specifications

Quantity 200 L
Molecular Weight (g/mol) 40.00
Percent Purity 48 to 52%
Grade Reagent
Identification To Pass Test
Packaging Drum
Chemical Name or Material Sodium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.