missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/w), Reagents

Catalog No. 01333555
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01333555 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333555 Supplier Labchem Inc Supplier No. CS1681001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Hydroxide, 50% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH >13
Color Colorless
Boiling Point 140°C
Quantity 1 L
Percent Purity 50%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.5267
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 50% (w/w)