missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% w/w Reagent (760 g/L), Reagents

Catalog No. 01337724
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337724 10 L
01337723 1 L
01337722 200 L
01337721 20 L
01337720 4 L
01337719 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01337724 Supplier Labchem Inc Supplier No. C131860010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Hydroxide, 50% w/w Reagent (760 g/L), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH >13
Color Colorless
Boiling Point 140°C
Melting Point 12°C
Quantity 10 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.5267
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 50% w/w Reagent (760 g/L)