missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/w) (760 g/L), Analytical Reagent Grade, Ricca Chemical

Catalog No. p-2917128
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
30 gal.
Packaging:
Hard Pack
Natural Poly Bottle
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
729116 500 mL Natural Poly Bottle
729132 1 L Natural Poly Bottle
72911 4 L Natural Poly Bottle
72915HP 20 L Hard Pack
729130 30 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 729116 Supplier Ricca Chemical Company Supplier No. 729116
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

CAS Min % 49.0
CAS Max % 51.0
Quantity 500 mL
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
Percent Purity 50 to 52%
Grade Analytical
Concentration or Composition (by Analyte or Components) 50% (w/w) Aqueous Solution, 760g/L
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide