missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/w) (760 g/L), Analytical Reagent Grade, Ricca Chemical

Supplier:  Ricca Chemical Company 72915HP

 View more versions of this product

Catalog No. 72915HP


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide
49,49
HNaO
20 L
[OH-].[Na+]
39.997
CHEBI:32145
Analytical
Hard Pack
1310-73-2,7732-18-5
51,51
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
50 to 52%
50% (w/w) Aqueous Solution, 760g/L

Chemical Identifiers

1310-73-2
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
[OH-].[Na+]
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions