missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/w) (760 g/L), Purified Grade, Ricca Chemical

Nitrogen Free - Suitable for Kjeldahl Nitrogen Analysis

$123.17 - $1199.27

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
729016
SDS View Documents
Ricca Chemical Company
729016
500 mL Natural Poly Bottle
Each for $123.17
Only null left
 
729032
View Documents
Ricca Chemical Company
729032
1 L Natural Poly Bottle
Each for $175.13
Only null left
 
7290-1
SDS Accessories View Documents
Ricca Chemical Company
72901
4 L Natural Poly Bottle
Each for $406.38
Only null left
 
7290-5
SDS View Documents
Ricca Chemical Company
72905
20 L Cubitainer
Each for $1,199.27
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1310-73-2
HEMHJVSKTPXQMS-UHFFFAOYSA-M
CHEBI:32145
HNaO
14798
[OH-].[Na+]

Specifications

Colorless
49.0,49.0
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
min 49.5% w/w
Natural Poly Bottle
Liquid
1310-73-2,7732-18-5
51.0,51.0
50% (w/w), 760 g/L
500 mL
[OH-].[Na+]
39.997
CHEBI:32145
Purified
Aqueous Solution
Sodium Hydroxide
Videos
Safety and Handling

Safety and Handling

ShelfLife : 36 months

SDS
missing translation for 'documents'

missing translation for 'documents'