missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/w) (760 g/L), Purified Grade, Ricca Chemical

Catalog No. 729016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
729016 500 mL Natural Poly Bottle
729032 1 L Natural Poly Bottle
72901 4 L Natural Poly Bottle
72905 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 729016 Supplier Ricca Chemical Company Supplier No. 729016
Only null left

Nitrogen Free - Suitable for Kjeldahl Nitrogen Analysis

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 49.0
CAS Max % 51.0
Concentration or Composition Notes 50% (w/w), 760 g/L
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Quantity 500 mL
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
Percent Purity min 49.5% w/w
Grade Purified
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
ShelfLife 36 months