missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/v) Solution, Spectrum™ Chemical

Catalog No. 18613613
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613613 4 L Poly Bottle
18613665 500 mL Poly Bottle
18613612 1 L Poly Bottle
18613614 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613613 Supplier Spectrum Chemical Mfg Cor Supplier No. S2244LTPL
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5

Specifications

Quantity 4 L
Percent Purity 50% ±1%
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Sodium Hydroxide Solution

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.