missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 50% w/v, Reagents
Specifications
Specifications
| pH | 14 |
| Color | Colorless |
| Boiling Point | 112°C |
| Quantity | 4 L |
| Percent Purity | 50% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.35 |
| Physical Form | Liquid |
| Chemical Name or Material | Sodium Hydroxide, 50% w/v |