missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 50% (w/v), Analytical Reagent Grade, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 49.0, 48.61 |
| CAS Max % | 51.0, 50.6 |
| Concentration or Composition Notes | 50% (w/v) |
| Quantity | 500 mL |
| IUPAC Name | sodium;hydroxide |
| Molecular Weight (g/mol) | 39.997 |
| Percent Purity | 49.92 to 50.08% |
| Grade | Analytical |
| Packaging | Natural Poly Bottle |
| Show More |