missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/v), Analytical Reagent Grade, Ricca Chemical

Catalog No. 72835
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
72835 20 L Cubitainer
728316 500 mL Natural Poly Bottle
728332 1 L Natural Poly Bottle
72831 4 L Natural Poly Bottle
728325 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 72835 Supplier Ricca Chemical Company Supplier No. 72835

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for Orsat Gas Analysis

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Quantity 20 L
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Percent Purity 49.92 to 50.08%
Grade Analytical
Concentration or Composition (by Analyte or Components) 50% (w/v) Aqueous Solution
Packaging Cubitainer
Chemical Name or Material Sodium Hydroxide