missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 5% (w/v) (50 g/L), Ricca Chemical

Catalog No. 725016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
725016 500 mL Natural Poly Bottle
725032 1 L Natural Poly Bottle
72501 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 725016 Supplier Ricca Chemical Company Supplier No. 725016

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 500 mL
Percent Purity 4.96 to 5.04%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide