missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 5.21 N (17.5% w/w), Ricca Chemical
Supplier: Ricca Chemical Company R74652101A
| Quantity | 1 L |
|---|---|
| Packaging | Natural Poly Bottle |
Chemical Identifiers
| 1310-73-2 , 7732-18-5 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Specifications
| Sodium Hydroxide | |
| 69.0, 28.9 | |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| Natural Poly Bottle | |
| 5.21Normal,17.5% (w/w) |