missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 5.0 Normal, Reagents

Catalog No. 01333847
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333847 1 L
01333848 10 L
01333846 20 L
01333845 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01333847 Supplier Labchem Inc Supplier No. CS1040001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Hydroxide, 5.0 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH >13
Color Colorless
Boiling Point 100°C
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1873
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 5.0 Normal