missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 40% (w/w), Analytical Reagent Grade, Ricca Chemical

Catalog No. 72885
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
72885 20 L Cubitainer
728832 1 L Natural Poly Bottle
72881 4 L Natural Poly Bottle
728825 10 L Cubitainer
728816 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 72885 Supplier Ricca Chemical Company Supplier No. 72885

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Kjeldahl Nitrogen analysis

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Quantity 20 L
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Percent Purity 39.8 to 40.2%
Grade Analytical
Concentration or Composition (by Analyte or Components) 40% (w/w) Aqueous Solution
Packaging Cubitainer
Chemical Name or Material Sodium Hydroxide, Nitrogen Free