missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 40% w/v aq. soln.

CAS: 1310-73-2 | HNaO | 39.997 g/mol

$208.30 - $448.38

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35639K7
View Documents
Thermo Scientific Chemicals
035639K7
4 L
Each for $208.30
Only null left
 
AA35639M4
View Documents
Thermo Scientific Chemicals
035639M4
20 L
Each for $448.38
Only null left
 
Description

Description

Sodium hydroxide, 40% w/v aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing of food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]

Specifications

1310-73-2
MFCD00003548
40% w/v aq. soln.
Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether.
[OH-].[Na+]
39.997
CHEBI:32145
4 L
Sodium hydroxide
HNaO
UN1824
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE SOLUTION

EINECSNumber : 215-185-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only