missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 4% (w/v) (40 g/L), Ricca Chemical

Supplier:  Ricca Chemical Company 724716

 View more versions of this product

Catalog No. 724716


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide
1310-73-2,7732-18-5
98.0,3.93
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Laboratory
Aqueous Solution
500 mL
4% (w/v),40g/L
Colorless
94.0,3.77
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
4%
Natural Poly Bottle
>13
Liquid

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions