missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 32% w/v, Reagents

Catalog No. 01333736
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333736 1 L
01333735 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333736 Supplier Labchem Inc Supplier No. CS1301001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Hydroxide, 32% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH >13
Color Colorless
Boiling Point 100°C
Quantity 1 L
Percent Purity 32%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.2776
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 32% w/v