missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 30% w/v, Reagents

Catalog No. 01333833
Change view
Click to view available options
Quantity:
4 L
20 L
15 gal.
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333833 4 L
01333836 15 gal.
01333835 200 L
01333834 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01333833 Supplier Labchem Inc Supplier No. CS1045004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Hydroxide, 30% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH >13
Color Colorless
Boiling Point 108°C
Quantity 4 L
Percent Purity 30%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.2627
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 30% w/v