missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 20% (w/v) (200 g/L), Ricca Chemical

Catalog No. p-3148259
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
727016 500 mL Natural Poly Bottle
727032 1 L Natural Poly Bottle
72701 4 L Natural Poly Bottle
13622616 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 727016 Supplier Ricca Chemical Company Supplier No. 727016
Only null left

P4TF8-S12, Class F

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Min % 82.0, 16.34
CAS Max % 85.0, 17.01
Quantity 500 mL
Percent Purity 19.96 to 20.04%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 20% (w/v) Aqueous Solution (200g/L)
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide