missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 20°C Baumé, Reagent Grade, Ricca Chemical

Catalog No. p-4770416
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R7265000500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R7265000500 Supplier Ricca Chemical Company Supplier No. R7265000500A
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Assay Percent Range 14.36 ± 0.14% (w/w)
Solubility Information Solubility in Water: Infinite
Quantity 500 mL
Percent Purity 14.22 to 14.5%
Grade Reagent
Odor Odorless
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 20°Baume
Show More Show Less