missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 2 N Aqueous Solution, Spectrum™ Chemical

Catalog No. 18613583
Change view
Click to view available options
Quantity:
1 L
4 L
200 L
Packaging:
Poly Bottle
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613583 1 L Poly Bottle
18613584 4 L Poly Bottle
18613582 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613583 Supplier Spectrum Chemical Mfg Cor Supplier No. S3751LT
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5

Specifications

Concentration 2 N ± 0.03 N
Quantity 1 L
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Sodium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.