missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 2.0N Standardized Solution

CAS: 1310-73-2 | HNaO | 39.997 g/mol

$89.15 - $169.82

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35631K2
View Documents
Thermo Scientific Chemicals
035631K2
1 L
Each for $89.15
Only null left
 
AA35631K7
View Documents
Thermo Scientific Chemicals
035631K7
4 L
Each for $169.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
CHEBI:32145
[OH-].[Na+]
HNaO
MFCD00003548
14798
sodium;hydroxide

Specifications

1310-73-2
Liquid
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
1 L
CHEBI:32145
Sodium hydroxide
Odorless
HNaO
Colorless
[OH-].[Na+]
39.997
14798
40
Videos
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 215-185-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only