missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 2.00 N, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 91.0, 7.4 |
| CAS Max % | 94.0, 7.7 |
| Concentration or Composition Notes | 2.00Normal |
| Quantity | 4 L |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Physical Form | Liquid |
| Chemical Name or Material | Sodium Hydroxide |