missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 15% w/w, Reagents
Specifications
Specifications
| pH | 14 |
| Color | Colorless |
| Boiling Point | 104°C |
| Quantity | 20 L |
| Percent Purity | 15% |
| Grade | Reagent |
| Packaging | Cubitainer |
| Specific Gravity | 1.1661 |
| Physical Form | Liquid |
| Chemical Name or Material | Sodium Hydroxide, 15% w/w |