missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 15% w/v, Reagents

Catalog No. 01333565
Change view
Click to view available options
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01333565 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333565 Supplier Labchem Inc Supplier No. CS16050020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Hydroxide, 15% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH 14
Color Colorless
Boiling Point 104°C
Quantity 20 L
Percent Purity 15%
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.145
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 15% w/v