missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide, 10.0N Standardized Solution
Description
Sodium hydroxide, 10.0N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Color | Colorless |
| UN Number | UN1824 |
| Concentration | 10.0N |
| Density | 1.327 g/mL |
| Solubility Information | Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether. |
| Formula Weight | 40 |
| Odor | Odorless |
| Quantity | 1 L |
| Physical Form | Liquid |
| Chemical Name or Material | Sodium hydroxide, Standardized Solution |
Sécurité et manipulation
| Hazard Category | H290-H314-H335 |
| Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
| Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE SOLUTION |
| EINECSNumber | 215-185-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only