missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 10.0 N, Ricca Chemical
$113.50 - $1012.50
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 39.997 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
747016
|
Ricca Chemical Company
747016 |
500 mL | Natural Poly Bottle |
Each for $113.50
|
|
||||
|
7470-32
|
Ricca Chemical Company
747032 |
1 L | Natural Poly Bottle |
Each for $165.33
|
|
||||
|
R74700004F
|
Ricca Chemical Company
R74700004F |
4 L | Cubitainer |
Case for $283.70
|
|
||||
|
7470-1
|
Ricca Chemical Company
74701 |
4 L | Natural Poly Bottle |
Each for $371.23
|
|
||||
|
747025
|
Ricca Chemical Company
747025 |
10 L | Cubitainer |
Each for $711.13
|
|
||||
|
7470-5
|
Ricca Chemical Company
74705 |
20 L | Cubitainer |
Each for $1,012.50
|
|
||||
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Specifications
| 1310-73-2,7732-18-5 | |
| 57.0,45.43 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| Laboratory | |
| Natural Poly Bottle |
| 54.0,43.65 | |
| HNaO | |
| 500 mL | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| 10.0N | |
| Sodium Hydroxide |