missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 10.0 N, Ricca Chemical

Supplier:  Ricca Chemical Company 74705

 View more versions of this product

Catalog No. 7470-5


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide
54.0,43.65
HNaO
20 L
[OH-].[Na+]
39.997
CHEBI:32145
10.0N
1310-73-2,7732-18-5
57.0,45.43
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Laboratory
Cubitainer

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions