missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 1% (w/v) (10 g/L), Ricca Chemical

Supplier:  Ricca Chemical Company 72431

 View more versions of this product

Catalog No. 72431


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide Solution
97.0,0.97
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
1%
Natural Poly Bottle
>13
1% (w/v),10g/L
1310-73-2,7732-18-5
100.0,1.01
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Laboratory
Aqueous Solution
4 L

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Accessories
Promotions

Promotions