missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 1.00 N, Ricca Chemical
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Specifications
Specifications
| pH | >13 |
| Color | Colorless |
| Boiling Point | ∼100°C |
| Melting Point | ∼0°C |
| CAS Min % | 96.18, 3.82 |
| CAS Max % | 96.18, 3.82 |
| Density | 1.044 |
| Solubility Information | Miscible in water |
| Quantity | 1 L |
| Grade | Laboratory |
| Show More |
Safety and Handling
| ShelfLife | 24 months |
| Recommended Storage | Ambient Corrosive |