missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 1.00 N, Ricca Chemical

Catalog No. 745016
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
745016 500 mL Natural Poly Bottle
74504 120 mL Natural Poly Bottle
R74500001F 1 L Cubitainer
745032 1 L Natural Poly Bottle
74501CT 4 L Cubitainer
74501 4 L Natural Poly Bottle
745025 10 L Cubitainer
74505 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 745016 Supplier Ricca Chemical Company Supplier No. 745016
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Standardized Bases

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40 g/mol
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Min % 91%, 7.25
CAS Max % 94%, 7.54
Quantity 500 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1.00N
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide